missing translation for 'onlineSavingsMsg'
Learn More

DGK-alpha Antibody [Janelia Fluor« 549], Novus Biologicals Biologicals™

Codice prodotto. 30510416 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
Dimensione della confezione:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30510416 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30510416 Supplier Novus Biologicals Supplier No. NBP338348JF549

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DGK-alpha Polyclonal antibody specifically detects DGK-alpha in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigene DGK-alpha
Applicazioni ELISA, Western Blot
Classificazione Polyclonal
Coniugato Janelia Fluor 549
Formulazione 50mM Sodium Borate
Alias gene DAG kinase alpha, DAGK, DAGK1diacylglycerol kinase, alpha (80kD), DGK-alphaEC 2.7.1.107, diacylglycerol kinase alpha, diacylglycerol kinase, alpha 80kDa, Diglyceride kinase alpha, MGC12821,80 kDa diacylglycerol kinase, MGC42356
Specie ospite Rabbit
Immunogeno Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human DGK-alpha (NP_958852.1).,, Sequence:, MAKERGLISPSDFAQLQKYMEYSTKKVSDVLKLFEDGEMAKYVQGDAIGYEGFQQFLKIYLEVDNVPRHLSLALFQSFETGHCLNETNVTKDVVCLNDVSCYFSLLEGGRPEDKLEFTFKLYDTDRNGILDSSEVDKIILQMMRVAEYLDWDVSELRPILQEMMKEIDYDGSGSVSQAEWVRAGATTVPLLVLLGLEMTL
Metodo di purificazione Affinity purified
Quantità 0.1 mL
Status giuridico RUO
Disciplina di ricerca Lipid and Metabolism
Primario o secondario Primary
ID gene (immissione) 1606
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Forma Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.