missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Di-n-heptyl phthalate, 98+%

Marca:  Alfa Aesar™ A15466.18

Dettagli aggiuntivi : CAS : 3648-21-3 Peso : 0.05000kg

Quantità 50g

 vedi altre versioni di questo prodotto

Codice prodotto. 11402837

  • 182.00€ / 50g


Spedizione stimata
21-04-2021
Aggiungi al carrello

Descrizione

Descrizione

Applications
Di-n-heptyl Phthalate is a Phthalate derivative and an organic extract contaminant found in drinking water, which was shown to activate Nrf-2-Mediated Antioxidant response in human cell line.

Solubility
Sparingly soluble in water.

Notes
Incompatible with Strong acids, strong bases,.
Specifica

Identificatori chimici

3648-21-3
362.51
JQCXWCOOWVGKMT-UHFFFAOYSA-N
19284
diheptyl benzene-1,2-dicarboxylate
C22H34O4
MFCD00043680
diheptyl phthalate, di-n-heptyl phthalate, heptyl phthalate, di-n-heptylphthalate, phthalic acid, diheptyl ester, phthalic acid diheptyl ester, 1,2-benzenedicarboxylic acid, diheptyl ester, n-diheptyl phthalate, unii-ie05vo8p8p, diheptyl ester of phthalic acid
CHEBI:34677
CCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCC

Specifica

Di-n-heptyl phthalate
213°C (415°F)
360°C
3648-21-3
MFCD00043680
diheptyl phthalate, di-n-heptyl phthalate, heptyl phthalate, di-n-heptylphthalate, phthalic acid, diheptyl ester, phthalic acid diheptyl ester, 1,2-benzenedicarboxylic acid, diheptyl ester, n-diheptyl phthalate, unii-ie05vo8p8p, diheptyl ester of phthalic acid
JQCXWCOOWVGKMT-UHFFFAOYSA-N
diheptyl benzene-1,2-dicarboxylate
19284
362.51
0.992
1.486
50g
C22H34O4
2336623
Sparingly soluble in water.
CCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCC
362.51
CHEBI:34677
≥98%
Videos
SDS Documenti