missing translation for 'onlineSavingsMsg'
Learn More

Diacetone-D-glucose, 98+%, Thermo Scientific Chemicals

Catalog Number p-7022681
Click to view available options
Quantità:
25 g
100 g
500 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
582-52-5
C12H20O6
260.286
MFCD00005544
KEJGAYKWRDILTF-MQIGXGKASA-N
1,2:5,6-diisopropylidene-d-glucose, 1,2:5,6-di-o-isopropylidene-alpha-d-glucofuranose, 3ar,5r,6s,6as-5-4s-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol
40469256
(3aR,5S,6S,6aS)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
582-52-5
C12H20O6
260.286
MFCD00005544
KEJGAYKWRDILTF-MQIGXGKASA-N
1,2:5,6-diisopropylidene-d-glucose, 1,2:5,6-di-o-isopropylidene-alpha-d-glucofuranose, 3ar,5r,6s,6as-5-4s-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol
40469256
(3aR,5S,6S,6aS)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C

It is mainly used in biochemical reaction and used as medicine intermediate. It is used in synthesize for below products: L-gulose;1,2:5,6-Di-O-isopropylidene-a-D-gulofuranose; Alllose; ;1,2:5,6-Double-O-Isopropylidene-3-C-(1-Methoxycarbonyl)Ethide-alfa-D-Furanose) ;1,2:5,6-Di-O-isopropylidene-a-D-ribo-hexofuranose-3-ulose;6-deoxy idose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is mainly used in biochemical reaction and used as medicine intermediate. It is used in synthesize for below products: L-gulose;1,2:5,6-Di-O-isopropylidene-a-D-gulofuranose; Alllose; ;1,2:5,6-Double-O-Isopropylidene-3-C-(1-Methoxycarbonyl)Ethide-alfa-D-Furanose) ;1,2:5,6-Di-O-isopropylidene-a-D-ribo-hexofuranose-3-ulose;6-deoxy idose.

Solubility
Solubility in hot water gives very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Diacetone-D-glucose
Punto di fusione 107°C to 110°C
Quantità 25 g
Beilstein 84386
Indice di Merck 14,2965
Optical Rotation −11° (c=5 in Ethanol)
Peso formulazione 260.29
Percent Purity ≥98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'