Learn More
Diallyl maleate, 95%
Codice prodotto. 11366686
Cambia vista
Click to view available options
Quantità:
50 g
250 g
1000 g
Dimensione della confezione:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
11366686 250 g 250g
11356686 50 g 50g
11376686 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 11366686 Fornitore Thermo Scientific Alfa Aesar N. del fornitore L03867.30
Richiesta di formato massivo o personalizzato
CAS: 999-21-3 | C10H12O4 | 196.202 g/mol
Diallyl maleate has potential applications in polyester resins, adhesives, and ion exchange resins. When used at low levels, Diallyl Maleate is an effective agent for the promotion of branching in emulsion polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diallyl maleate has potential applications in polyester resins, adhesives, and ion exchange resins. When used at low levels, Diallyl Maleate is an effective agent for the promotion of branching in emulsion polymers.
Solubility
Soluble in water 151.2 mg/L @ 25°C.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 999-21-3
Formula molecolare C10H12O4
Molecular Weight (g/mol) 196.202
Numero MDL MFCD00010763
InChI Key ZPOLOEWJWXZUSP-WAYWQWQTSA-N
Sinonimo diallyl fumarate, fumaric acid, diallyl ester, unii-k852lk4c5p, 2-butenedioic acid e-, di-2-propenyl ester, 2-butenedioic acid 2e-, di-2-propenyl ester, butenedioic acid diallyl ester, 1,4-bis prop-2-en-1-yl 2e-but-2-enedioate, 2-butenedioic acid 2e-, 1,4-di-2-propen-1-yl ester, diallyl malleate, 1,4-bis prop-2-en-1-yl but-2-enedioate
PubChem CID 5354266
IUPAC Name bis(prop-2-enyl) (Z)-but-2-enedioate
SMILES C=CCOC(=O)C=CC(=O)OCC=C
Specifica
Materiale o nome chimico Diallyl maleate
Punto di fusione -47°C
Densità 1.072
Punti di ebollizione 109°C to 110°C (3 mmHg)
Punto d'infiammabilità 130°C (266°F)
Indice di rifrazione 1.467
Quantità 250 g
Numero UN UN2810
Beilstein 1725954
Informazioni di solubilità Soluble in water 151.2mg/L @ 25°C.
Peso formulazione 196.21
Percent Purity 95%