Learn More
Sartorius Stedim Biotech™ Diatomaceous Earth Filter, Discs, Grade 470
Made of cellulose and diatomaceous earth with much better separating capability than pure cellulose papers at the same rate of filtration.
Marca:  Sartorius Stedim Biotech™ FT-3-606-110
Dettagli aggiuntivi : Peso : 0.16000kg
Codice prodotto. 15922970
  • 55.30€ / pezzi 100
Spedizione stimata: 03-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Quickly retains the finest particles at high flow rates.
  • Suitable for the following:
    • Clarification of beer, wine, urine during spectrophotometric or refractometric tests.
    • Filtration of the finest, semi-colloidal precipitates, e.g., those consisting of proteins, clay or cold-precipitated barium.
    • Analytical sample preparation.
  • Non-sterile, bulk packed.
Specifica
Specifica
140 g/m2
Cellulose, Diatomaceous Earth
470
0.32 mm
Disc
80 sec.
100/Pk.
110 mm