missing translation for 'onlineSavingsMsg'
Learn More

Diazoxon, TRC

Codice prodotto. 18022550
Cambia vista
Click to view available options
Quantità:
100 mg
250 mg
1 g
Dimensione della confezione:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18022550 100 mg 100mg
18001150 250 mg 250mg
18003840 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18022550 Fornitore Toronto Research Chemicals N. del fornitore TRCD416890100MG

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 962-58-3
Formula molecolare C12 H21 N2 O4 P
Molecular Weight (g/mol) 288.28
Sinonimo Phosphoric acid, diethyl 6-methyl-2-(1-methylethyl)-4-pyrimidinyl ester, Phosphoric acid, diethyl 2-isopropyl-6-methyl-4-pyrimidinyl ester (6CI,7CI,8CI), 4-Pyrimidinol, 2-isopropyl-6-methyl-, diethyl phosphate (6CI), Diazinon oxon, Diazoxon, Diethyl 2-isopropyl-6-methyl-4-pyrimidinyl phosphate, O,O-Diethyl O-(2-isopropyl-6-methyl-4-pyrimidinyl) phosphate, O,O-Diethyl-O-(2-isopropyl-4-methylpyrimidyl-6) phosphate, Oxodiazinon
IUPAC Name diethyl (6-methyl-2-propan-2-ylpyrimidin-4-yl) phosphate
SMILES CCOP(=O)(OCC)Oc1cc(C)nc(n1)C(C)C

Specifications

Materiale o nome chimico Diazoxon
Note sul grado di purezza HPLC
Quantità 100 mg
InChI Formula InChI=1S/C12H21N2O4P/c1-6-16-19(15,17-7-2)18-11-8-10(5)13-12(14-11)9(3)4/h8-9H,6-7H2,1-5H3
Peso formulazione 288.1239
Percent Purity >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.