Learn More
Dibenzothiophene 5-Oxide, TRC
Product Code. 30379271
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30379271 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30379271 Supplier Toronto Research Chemicals Supplier No. TRCD42312550MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dibenzothiophene 5-Oxide
CAS 1013-23-6
Molecular Formula C12H8OS
Quantity 50 mg
InChI Formula InChI=1S/C12H8OS/c13-14-11-7-3-1-5-9(11)10-6-2-4-8-12(10)14/h1-8H
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3S2=O
Molecular Weight (g/mol) 200.25
Formula Weight 200.0296