Learn More
Dibenzyl Dicarbonate, TRC
Specifica
Specifica
| Materiale o nome chimico | Dibenzyl Dicarbonate |
| CAS | 31139-36-3 |
| Formula molecolare | C16H14O5 |
| Quantità | 5 g |
| Sinonimo | Dicarbonic Acid 1,3-Bis(phenylmethyl) Ester, Dicarbonic Acid Bis(phenylmethyl) Ester, Oxydiformic Acid Dibenzyl Ester, Dibenzyl Pyrocarbonate, Pyrocarbonic Acid Dibenzyl Ester |
| InChI Formula | InChI=1S/C16H14O5/c17-15(19-11-13-7-3-1-4-8-13)21-16(18)20-12-14-9-5-2-6-10-14/h1-10H,11-12H2 |
| SMILES | O=C(OCc1ccccc1)OC(=O)OCc2ccccc2 |
| IUPAC Name | benzyl phenylmethoxycarbonyl carbonate |
| Molecular Weight (g/mol) | 286.28 |
| Peso formulazione | 286.0841 |