missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha'-Dibromo-p-xylene, 97%, Thermo Scientific Chemicals

Catalog Number p-102095
46.00 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.


Click to view available options
Quantità:
25 g
100 g
Imballaggio:
Glass bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
623-24-5
C8H8Br2
263.95
MFCD00000182
RBZMSGOBSOCYHR-UHFFFAOYSA-N
1,4-bis bromomethyl benzene, p-xylylene dibromide, alpha,alpha'-dibromo-p-xylene, benzene, 1,4-bis bromomethyl, p-xylylene bromide, p-bromomethyl benzene, 1,4-dibromomethylbenzene, p-bis bromomethyl benzene, 1,4-bis-bromomethyl-benzene, ccris 1775
69335
1,4-bis(bromomethyl)benzene
C1=CC(=CC=C1CBr)CBr
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
623-24-5
C8H8Br2
263.95
MFCD00000182
RBZMSGOBSOCYHR-UHFFFAOYSA-N
1,4-bis bromomethyl benzene, p-xylylene dibromide, alpha,alpha'-dibromo-p-xylene, benzene, 1,4-bis bromomethyl, p-xylylene bromide, p-bromomethyl benzene, 1,4-dibromomethylbenzene, p-bis bromomethyl benzene, 1,4-bis-bromomethyl-benzene, ccris 1775
69335
1,4-bis(bromomethyl)benzene
C1=CC(=CC=C1CBr)CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico α,α′-Dibromo-p-xylene
Nota nome 97%
CAS min. % 96.0
CAS max % 100.0
Punto di fusione 142.0°C to 147.0°C
Colore White to Beige
Densità 2.0120g/mL
Punti di ebollizione 245.0°C
Spettro a infrarossi Authentic
Intervallo percentuale saggio 96% min. (GC)
Imballaggio Glass bottle
Formula lineare C6H4(CH2Br)2
Quantità 25 g
Beilstein 05, 385
Gravità specifica 2.012
Informazioni di solubilità Solubility in water: soluble,hydrolyses
Peso formulazione 263.95
Percent Purity 97%
Forma fisica Crystalline Powder and Coarse Crystals
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'