Learn More
Dibutyl Sebacate, TRC
Product Code. 30133003
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133003 5 g 5g
17970065 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30133003 Supplier Toronto Research Chemicals Supplier No. TRCD4295255G
 Restricted Item
Spedizione stimata: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 109-43-3
Molecular Formula C18 H34 O4
Molecular Weight (g/mol) 314.46
Synonym Decanedioic acid, dibutyl ester (9CI), Sebacic acid, dibutyl ester (6CI,8CI), Bis(n-butyl) sebacate, DBS, Di-n-Butyl sebacate, Dibutyl decanedioate, Dibutyl sebacate, Ergoplast SDB, Kodaflex DBS, NSC 3893, PX 404, Polycizer DBS, Reomol DBS, Sebacic acid di-n-butyl ester, Staflex DBS, Uniflex DBS
IUPAC Name dibutyl decanedioate
SMILES CCCCOC(=O)CCCCCCCCC(=O)OCCCC
Specifications
Quantity 5 g
InChI Formula InChI=1S/C18H34O4/c1-3-5-15-21-17(19)13-11-9-7-8-10-12-14-18(20)22-16-6-4-2/h3-16H2,1-2H3
Formula Weight 314.2457
Chemical Name or Material Dibutyl Sebacate