Product Code. 16358186
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
10 mL
25 mL
50 mL
Unit Size:
10mL
25mL
50mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
16358186
50 mL
Glass Bottle
50mL
10002260
5 mL
Glass Bottle
5mL
16348186
10 mL
Glass Bottle
10mL
10134290
25 mL
Glass Bottle
25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
16358186
Supplier
Thermo Scientific Acros
Supplier No.
176110500
830.00€ 760.00€ / 50mL Risparmia 70.00€ 8% Sconto
Spedizione stimata: 30-11-2026Eseguire il login per visualizzare lo stock disponibile
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
CAS: 1665-00-5 | CH2Cl2 | 86.939 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
Chemical Identifiers
CAS
1665-00-5
Molecular Formula
CH2Cl2
MDL Number
MFCD00000882
InChI Key
YMWUJEATGCHHMB-DICFDUPASA-N
Synonym
dichloromethane-d2, 99.9 atom % d, dichloromethane-d2, dichloro dideuterio methane, cd2cl2, dichloromethane-d', 99.96 atom % d, dichloro 2h2 methane, dideuteromethylene chloride, methylene chloride-d2, methane-d2, dichloro, deuterated dichloromethane
PubChem CID
160586
Specifications
Physical Form
Liquid
Color
Colorless
Density
1.3600 g/mL
Deuterium (D)
99.60% min.
Packaging
Glass Bottle
Refractive Index
1.4218
Water
0.04% max. (Coulometric)
Quantity
50 mL
Melting Point
-97.0°C
Boiling Point
40.0°C
Linear Formula
CD2 Cl2
Beilstein
01, IV, 39
SMILES
[2H]C([2H])(Cl)Cl
IUPAC Name
dichloro(²H₂)methane
Molecular Weight (g/mol)
86.94
Formula Weight
86.95
Grade
NMR
Chemical Name or Material
Dichloromethane-d2, 99.6+ atom % D
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?