Learn More
Didemethyl Mifepristone, TRC
Product Code. 30011086
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011086 1 mg 1mg
30133946 10 mg 10mg
30010135 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011086 Supplier Toronto Research Chemicals Supplier No. TRCD4395501MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 104004-92-4
Molecular Formula C27 H31 N O2
Molecular Weight (g/mol) 401.54
Synonym Estra-4,9-dien-3-one, 11-(4-aminophenyl)-17-hydroxy-17-(1-propyn-1-yl)-, (11β,17β)-, Estra-4,9-dien-3-one, 11-(4-aminophenyl)-17-hydroxy-17-(1-propynyl)-, (11β,17β)- (9CI), (11β,17β)-11-(4-Aminophenyl)-17-hydroxy-17-(1-propyn-1-yl)estra-4,9-dien-3-one, Aminopristone, Desmethylmetapristone, N,N-Didesmethylmifepristone, RU 42848, Didemethyl Mifepristone
IUPAC Name (8S,11R,13S,14S,17S)-11-(4-aminophenyl)-17-hydroxy-13-methyl-17-prop-1-ynyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
SMILES CC#C[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CCC4=C3[C@H](C[C@]12C)c5ccc(N)cc5
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C27H31NO2/c1-3-13-27(30)14-12-24-22-10-6-18-15-20(29)9-11-21(18)25(22)23(16-26(24,27)2)17-4-7-19(28)8-5-17/h4-5,7-8,15,22-24,30H,6,9-12,14,16,28H2,1-2H3/t22-,23+,24-,26-,27-/m0/s1
Formula Weight 401.2355
Percent Purity >95%
Chemical Name or Material Didemethyl Mifepristone
Grade HPLC