Learn More
Diethanolamine solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
BIOPHARMA
Product Code. 12972115 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12972115 500 mL Bottle 500mL
12952115 100 mL Bottle 100mL
12962115 250 mL Bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12972115 Supplier Fisher Chemical Supplier No. A 016 500
  • ONLINE EXCLUSIVE PRICE
    138.00€
    124.66€ / 500mL
    Risparmia 13.34€
    9% Sconto
 Restricted Item
Spedizione stimata: 28-12-2026
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Chemical Identifiers
CAS 111-42-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
MDL Number MFCD00002843
InChI Key ZBCBWPMODOFKDW-UHFFFAOYSA-N
PubChem CID 8113
ChEBI CHEBI:28123
IUPAC Name 2-(2-hydroxyethylamino)ethanol
SMILES OCCNCCO
Specifications
Packaging Bottle
Physical Form Liquid
Concentration 1 g/L
Type Single-component Standard Solution
Solvent or Matrix Water
Quantity 500 mL
Chemical Name or Material Diethanolamine