missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 10-undecyn-1-ylphosphonate, 95%, Thermo Scientific™

Catalog Number 15446057
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1242248-76-5
C15H29O3P
288.368
MFCD17012685
ZCOVBEAZCAHJDG-UHFFFAOYSA-N
diethyl 10-undecyn-1-ylphosphonate, diethyl undec-10-yn-1-ylphosphonate, 10-undecyn-1-ylphosphonic acid diethyl ester
97638951
11-diethoxyphosphorylundec-1-yne
CCOP(=O)(CCCCCCCCCC#C)OCC
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1242248-76-5
C15H29O3P
288.368
MFCD17012685
ZCOVBEAZCAHJDG-UHFFFAOYSA-N
diethyl 10-undecyn-1-ylphosphonate, diethyl undec-10-yn-1-ylphosphonate, 10-undecyn-1-ylphosphonic acid diethyl ester
97638951
11-diethoxyphosphorylundec-1-yne
CCOP(=O)(CCCCCCCCCC#C)OCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico Diethyl 10-undecyn-1-ylphosphonate
Formula lineare HC≡C(CH2)9P(O)(OCH2CH3)2
Indice di rifrazione 1.4460-1.4500
Quantità 1 g
Peso formulazione 288.37
Percent Purity 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'