Learn More
Diethyl Acetamidomalonate, Mikromol
Product Code. 30464283
Change view
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464283 4 x 25 mg -
30050075 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30464283 Supplier Mikromol Supplier No. MM3554.0300254AMP
 Restricted Item
Spedizione stimata: 30-11-2026
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Diethyl Acetamidomalonate, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Diethyl Acetamidomalonate
CAS 1068-90-2
Analyte or Component Names Diethyl Acetamidomalonate
Shipping Condition Room Temperature
Molecular Formula C9 H15 N O5
Certifications/Compliance ISO 17025
Synonym Propanedioic acid, 2-(acetylamino)-, 1,3-diethyl ester, Malonic acid, acetamido-, diethyl ester (6CI,7CI,8CI), Propanedioic acid, (acetylamino)-, diethyl ester (9CI), (Acetylamino)malonic acid diethyl ester, (Acetylamino)propanedioic acid diethyl ester, 2-(Acetylamino)propanedioic acid diethyl ester, 2-Acetylamino-malonic acid diethyl ester, Acetamidomalonic acid diethyl ester, Diethyl (N-acetylamino)malonate, Diethyl (acetylamino)malonate, Diethyl 2-(acetamido)propanedioate, Diethyl 2-(acetylamino)propanedioate, Diethyl 2-acetamidomalonate, Diethyl 2-acetylaminomalonate, Diethyl acetamidomalonate, Diethyl α-(acetylamino)malonate, NSC 7645
InChI Formula InChI=1S/C9H15NO5/c1-4-14-8(12)7(10-6(3)11)9(13)15-5-2/h7H,4-5H2,1-3H3,(H,10,11)
Solution Type Neat
SMILES CCOC(=O)C(NC(=O)C)C(=O)OCC
IUPAC Name diethyl 2-acetamidopropanedioate
Molecular Weight (g/mol) 217.22
Quantity 4 x 25 mg
Formula Weight 217.095 g/mol