Learn More
Diethyl carbonate, 99+%, Alfa Aesar™
Marca: Alfa Aesar A12477.0D
Dettagli aggiuntivi : CAS : 105-58-8 Peso : 0.01000kg Transport : UN number : 2366 Chem class : 3 Pack group : III
Quantità | 10000mL |
---|
Descrizione
ApplicationsDiethyl carbonate is esters, beta-enamino esters, carbamates and unsymmetrical alkyl carbonates. It is an active component of electrolytes used in lithium which is used as a solvent for cellulose ethers, nitro cellulose, natural and synthetic resin and in erythromycin intramuscular injections. It is also used in the synthesis of 3-ethyl-4-methyl-5-phenyl-3H-oxazol-2-one, phenobarbital, pyrethrum batteries.
Solubility
Miscible with diehtyl ether, ethanol and chloroform.Immiscible with water.
Notes
Store in cool place. Moisture sensitive. Incompatible with strong acids, oxidizing agents, strong bases and reducing agents.
Identificatori chimici
105-58-8 | |
118.132 | |
OIFBSDVPJOWBCH-UHFFFAOYSA-N | |
7766 | |
CCOC(=O)OCC |
C5H10O3 | |
MFCD00009107 | |
ethyl carbonate, carbonic acid, diethyl ester, carbonic acid diethyl ester, eufin, diatol, diaethylcarbonat, carbonic ether, ethoxyformic anhydride, diethylkarbonat, ethyl carbonate eto 2co | |
diethyl carbonate |
Specifica
Diethyl carbonate | |
31°C (87°F) | |
1.3845 | |
126°C to 128°C | |
10000mL | |
≥99% | |
MFCD00009107 | |
956591 | |
ethyl carbonate, carbonic acid, diethyl ester, carbonic acid diethyl ester, eufin, diatol, diaethylcarbonat, carbonic ether, ethoxyformic anhydride, diethylkarbonat, ethyl carbonate eto 2co | |
OIFBSDVPJOWBCH-UHFFFAOYSA-N | |
diethyl carbonate | |
7766 | |
Liquid |
0.975 | |
Pleasant | |
Moisture sensitive | |
-43°C | |
105-58-8 | |
C5H10O3 | |
UN2366 | |
14,3780 | |
Miscible with diehtyl ether,ethanol and chloroform.Immiscible with water. | |
CCOC(=O)OCC | |
118.132 | |
118.13 | |
≥99% |