Learn More
Alfa Aesar™ Diethyl formamidomalonate, 97%
Marca: Alfa Aesar™ L01189.14
Dettagli aggiuntivi : CAS : 6326-44-9 Peso : 0.02500kg
Quantità | 25g |
---|
Identificatori chimici
6326-44-9 | |
203.194 | |
PFLHGSJLYNJIOF-UHFFFAOYSA-N | |
80617 | |
CCOC(=O)C(C(=O)OCC)NC=O |
C8H13NO5 | |
MFCD00003279 | |
diethyl formamidomalonate, diethyl 2-formamidomalonate, diethyl 2-formylaminomalonate, diethyl formaminomalonate, diethyl2-formylaminomalonate, 1,3-diethyl 2-formamidopropanedioate, diethyl 2-carbonylaminopropane-1,3-dioate, formamidomalonic acid, diethyl ester, malonic acid, formamido-, diethyl ester, diethylformaminomalonate | |
diethyl 2-formamidopropanedioate |
Specifica
Diethyl formamidomalonate | |
50°C to 55°C | |
6326-44-9 | |
MFCD00003279 | |
diethyl formamidomalonate, diethyl 2-formamidomalonate, diethyl 2-formylaminomalonate, diethyl formaminomalonate, diethyl2-formylaminomalonate, 1,3-diethyl 2-formamidopropanedioate, diethyl 2-carbonylaminopropane-1,3-dioate, formamidomalonic acid, diethyl ester, malonic acid, formamido-, diethyl ester, diethylformaminomalonate | |
CCOC(=O)C(C(=O)OCC)NC=O | |
203.194 | |
203.19 |
173°C to 176°C (11mmHg) | |
25g | |
C8H13NO5 | |
1788903 | |
PFLHGSJLYNJIOF-UHFFFAOYSA-N | |
diethyl 2-formamidopropanedioate | |
80617 | |
97% |