missing translation for 'onlineSavingsMsg'
Learn More

Diethyl iminodiacetate, 97%, Thermo Scientific Chemicals

Catalog Number p-7040752
Click to view available options
Quantità:
5 mL
25 mL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6290-05-7
MFCD00041925
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6290-05-7
MFCD00041925

It is mainly used for the manufacture of N-(Phosphonomethyl) iminodiacetic acid and glyphosate. Used as raw material for ion exchange resin; for rubbers, electroplating and food additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is mainly used for the manufacture of N-(Phosphonomethyl) iminodiacetic acid and glyphosate. Used as raw material for ion exchange resin; for rubbers, electroplating and food additives.

Solubility
Moderately soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Diethyl iminodiacetate
Densità 1.056
Punti di ebollizione 208°C
Quantità 5 mL
Indice di rifrazione 1.435
Formula molecolare C8H10ClN
Formula lineare HN(CH2CO2C2H5)2
Sinonimo 1-4-chlorophenyl ethanamine, 1-4-chlorophenyl ethylamine, 4-chloro-alpha-methylbenzylamine, 1-4-chlorophenyl ethan-1-amine, 4-chloro-alpha-methyl benzylamine, 1-4-chlorophenyl-1-ethanamine, 1-4-chloro-phenyl-ethylamine, acmc-20amkz, acmc-20apje, acmc-209jma
Informazioni di solubilità Moderately soluble in water.
InChI Key PINPOEWMCLFRRB-UHFFFAOYNA-N
SMILES CC(C1=CC=C(C=C1)Cl)N
IUPAC Name 1-(4-chlorophenyl)ethanamine
Molecular Weight (g/mol) 155.63
PubChem CID 97803
Peso formulazione 189.21
Percent Purity 97%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'