Learn More
Diethyl Phthalate-d14, TRC
Specifica
Specifica
| Materiale o nome chimico | Phthalic acid diethyl ester d14 |
| CAS | 99873-99-1 |
| Formula molecolare | C12 2H14 O4 |
| Quantità | 5 mg |
| Sinonimo | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, di(ethyl-d5) ester (9CI), Phthalic acid diethyl ester D14, Phthalic acid, bis-ethyl ester D14, Diethyl phthalate-d14 |
| InChI Formula | InChI=1S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2,5D,6D,7D,8D |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)OC([2H])([2H])C([2H])([2H])[2H])c(C(=O)OC([2H])([2H])C([2H])([2H])[2H])c1[2H] |
| IUPAC Name | bis(1,1,2,2,2-pentadeuterioethyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 236.32 |
| Peso formulazione | 236.1771 |