Learn More
BD Aghi e anse per inoculazione monouso Difco™
Descrizione
Includes
Evans Blue Dye Certificate of Calibration with every box of 1,000 loops, confirming their accuracy and consistency
- Made from a unique plastic resin for controlled flexibility
- Resist cutting the agar surface, yet provide sufficient firmness to pick stubborn colonies
- Free of lubricants, oils and electrostatic charges
- Color coded
Specifica
Specifica
| Monouso | Sì |
| Volume | 10 μL |
| Materiale | Resina plastica |
| Sterilità | Sterile |
| Colore | Azzurro |
| Product Type | Anse per inoculazione |
| Quantità | 250/Pk. |