Learn More
Difenidol Hydrochloride, Mikromol™
Product Code. 17962706
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962706 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17962706 Supplier Mikromol Supplier No. MM3053.00
 Restricted Item
Spedizione stimata: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Difenidol Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Difenidol Hydrochloride
CAS 3254-89-5
Analyte or Component Names Difenidol Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C21 H27 N O . Cl H
Certifications/Compliance ISO 17034
Synonym Ansmin, Cefadol, Celmidol, Difenidol hydrochloride, Difenidolin, Diphenidol hydrochloride, Maniol, Mecalmin, Pineroro, SKF 478A, Satanolon, Tenesdol, Vontrol, Wansar, 1-Piperidinebutanol, alpha,alpha-diphenyl-, hydrochloride (1:1), 1-Piperidinebutanol, alpha,alpha-diphenyl-, hydrochloride (6CI,7CI,8CI,9CI), SKF-478A
InChI Formula InChI=1S/C21H27NO.ClH/c23-21(19-11-4-1-5-12-19,20-13-6-2-7-14-20)15-10-18-22-16-8-3-9-17-22;/h1-2,4-7,11-14,23H,3,8-10,15-18H2;1H
SMILES Cl.OC(CCCN1CCCCC1)(c2ccccc2)c3ccccc3
IUPAC Name 1,1-diphenyl-4-piperidin-1-ylbutan-1-ol;hydrochloride
Molecular Weight (g/mol) 345.91
Quantity 250 mg
Formula Weight 345.1859 g/mol