Learn More
Cole-Parmer™ Digi-Sense™ RTD Wire
Codice prodotto. 11795019 Sfoglia Tutto Cole-Parmer Prodotti
Cambia vista
Click to view available options
Temperature Range (Metric):
-200°C to +204°C
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Temperature Range (Metric) Dimensione della confezione
11795019 -200°C to +204°C Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11795019 Fornitore Cole-Parmer™ N. del fornitore 0854231
Spedizione stimata: 22-01-2027
Extend the measuring distance of your probes
  • Three-strand RTD wire features excellent resistance to abrasion, chemicals and moisture
  • Each silver-plated copper strad is color-coded with fluorinated ethylene propylene (FEP)
  • Three starnds are twisted to increase flexibility and minimize electromagnetic noise, then jacketed in white FEP
  • FEP insulation provides temperature resistance up to 400°F, and chemical resistance to solvents, acids and oils
  • Additionally, the insulation protects the wire from thermal aging, while maintaining its strength and flexibility
Specifica
Lunghezza (sistema metrico) 30.48 m
Materiale PTFE
Lunghezza (sistema imperiale) 100 ft.
Intervallo di temperatura (sistema imperiale) -328 to +400°F
Temperature Range (Metric) -200°C to +204°C