Learn More
Cole-Parmer™ Digi-Sense Traceable™ Color Frame Four-Channel Timer with Calibration
Sfoglia Tutto Cole-Parmer ProdottiDescrizione
Includes:
- Three colorful faceplates/frames, built-in stand/pocket clip, magnetic back, two 1.5 V button cell batteries and NIST-traceable calibration certificate with data.
- Three bright, colorful frames permit personalization, or dedicate a certain frame for a certain timing task
- Four independent timing channels feature unique audible alarms for simultaneous timing
- Count up or down from 100 hours for longer duration experiments
- Memory function returns the display to the programmed time
- Traceable products are calibrated to NIST-traceable standards and ready to use as soon as you receive it
- Each product goes through an intense vetting process before it can carry the Traceable name and it includes NIST-traceable certification, saving you both time and money by not having to calibrate separately
- NIST-traceable calibration also helps you meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirement
Specifica
Specifica
| Product Type | 4-channel Timer |
| Intervallo di tempo | 100 hr. |
| N. di canali | 4 |
| Certificazione/Conformità | NIST-Traceable |