Learn More
Cole-Parmer™ CC-200 Stuart™ Digital Colony Counter with IQ/OQ Documents
Codice prodotto. 15573449
Cambia vista
Click to view available options
Tensione:
90/230 VAC
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tensione Dimensione della confezione
15573449 90/230 VAC Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15573449 Fornitore Cole-Parmer™ N. del fornitore 1421206
  • ONLINE EXCLUSIVE PRICE
    1810.00€
    1571.08€ / pezzi 1
    Risparmia 238.92€
    13% Sconto
Visualizza tutte le promozioni
Spedizione stimata: 29-12-2026
The CC-200 Stuart™ Digital Colony Counter is a versatile unit can be used with 50 to 90 mm Petri dishes.
  • Compatible with felt tip markers for pressure-sensitive counting
  • Read individual plates or calculate an average over multiple plates
  • Energy-saving bright-white, LED light source
  • Antimicrobial coating provides additional protection
  • Audible count confirmation, adjustable pressure sensitivity, and option for light/dark background increases flexibility
  • Connects to a printer or computer to record individual or average plate counts
  • Includes IQ/OQ documents
  • 3 year warranty
  • Previous part number SC6PLUS/IQOQ
This colony counter registers a count each time you press a felt tip marker to the Petri dish. Cumulative count may be read on the digital display and is confirmed by an audible tone (can be turned on or off). Adjust the pressure required to measure a count — suitable to each user.
Sub-stage illumination is provided by energy-saving, bright LEDs for glare-free observation. Enhance counting of translucent or difficult-to-see colonies with the supplied black background filter. Calculate the average colony count over multiple plates using the averaging facility. Count results and statistics, including standard deviation, can be sent directly to a printer or computer using the supplied USB cable. Order the 1.7x and 3x magnification adapters separately for easier viewing of small colonies.
Please Note: Effective June 2022, the Stuart Digital Colony Counter Model number SC6PLUS/IQOQ has now been rebranded Cole-Parmer under Model number CC-200. All the same great features and specifications, just with a new and improved look.
Specifica
Product Type Colony Counter (Digital)
Dimensioni (L x P x A) 30 x 31 x 14 cm
Altezza (sistema imperiale) 5.5 in.
Altezza (sistema metrico) 14 cm
Lunghezza (sistema imperiale) 11.8 in.
Lunghezza (sistema metrico) 30 cm
Larghezza (sistema imperiale) 12.2 in.
Larghezza (sistema metrico) 31 cm
Frequenza 50/60 Hz
Tensione 90/230 VAC
Include Black Background Filter, Wolffhuegel Graticules (2), 50- to 90-mm Dish Centering Adapters (2), and UK/Shuko Power Plug Adapters