missing translation for 'onlineSavingsMsg'
Learn More

Diiodofluorescein

Codice prodotto. 11439220
Change view
Click to view available options
Quantità:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
11439220 50 g 50g
11429220 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439220 Supplier Thermo Scientific Alfa Aesar Supplier No. A16554.18
Richiesta di formato massivo o personalizzato

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

CAS: 31395-16-1 | C20H10I2O5 | 584.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31395-16-1
Formula molecolare C20H10I2O5
Molecular Weight (g/mol) 584.104
Numero MDL MFCD00036461
InChI Key RUEQYKGDINYYHI-UHFFFAOYSA-N
Sinonimo 4',5'-diiodofluorescein c.i. 45425:1, 3',6'-dihydroxy-4,5-diiodospiro 2-benzofuran-1,9'-xanthen-3-one
PubChem CID 21863873
IUPAC Name 3',6'-dihydroxy-6,7-diiodospiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC2=C(C=C1O)OC3=C(C24C5=C(C(=C(C=C5)I)I)C(=O)O4)C=CC(=C3)O

Specifications

Materiale o nome chimico Diiodofluorescein
Punto di fusione 239°C to 241°C (decomposition)
Quantità 50 g
Sensibilità Light sensitive
Peso formulazione 584.11

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.