missing translation for 'onlineSavingsMsg'
Learn More

(-)-Diisopropyl D-tartrate, 98%, Thermo Scientific Chemicals

Catalog Number p-7023617
Click to view available options
Quantità:
25 g
100 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
62961-64-2
C10H18O6
234.248
MFCD00008876
XEBCWEDRGPSHQH-YUMQZZPRSA-N
diisopropyl d-tartrate, 2s,3s-diisopropyl 2,3-dihydroxysuccinate, --diisopropyl d-tartrate, -diisopropyl-d-tartrate, d---tartaric acid diisopropyl ester, diisopropyl d---tartrate, butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, 2s,3s, d---diisopropyl tartrate, d-dipt, --d-tartaric acid diisopropyl ester, diisoprropyl-d-tartrate
112972
dipropan-2-yl (2S,3S)-2,3-dihydroxybutanedioate
CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
62961-64-2
C10H18O6
234.248
MFCD00008876
XEBCWEDRGPSHQH-YUMQZZPRSA-N
diisopropyl d-tartrate, 2s,3s-diisopropyl 2,3-dihydroxysuccinate, --diisopropyl d-tartrate, -diisopropyl-d-tartrate, d---tartaric acid diisopropyl ester, diisopropyl d---tartrate, butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, 2s,3s, d---diisopropyl tartrate, d-dipt, --d-tartaric acid diisopropyl ester, diisoprropyl-d-tartrate
112972
dipropan-2-yl (2S,3S)-2,3-dihydroxybutanedioate
CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico (-)-Diisopropyl D-tartrate
Densità 1.117
Punti di ebollizione 150°C to 151°C (11 mmHg)
Punto d'infiammabilità 110°C (230°F)
Indice di rifrazione 1.44
Quantità 25 g
Beilstein 5265431
Optical Rotation −16° (Neat)
Peso formulazione 234.25
Percent Purity 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'