missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals

Catalog Number 11394479
Click to view available options
Quantità:
10 g
100 g
500 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Normativo: Possono essere applicate restrizioni all'acquisto di questo prodotto e potremmo avere bisogno di richiedere ulteriore documentazione all'utente.
756-79-6
C3H9O3P
124.08
MFCD00008349
VONWDASPFIQPDY-UHFFFAOYSA-N
dimethyl methylphosphonate, dimethylmethylphosphonate, pyrol dmmp, dmmp, dimethyl methanephosphonate, fyrol dmmp, phosphonic acid, methyl-, dimethyl ester, methanephosphonic acid dimethyl ester, dimethoxymethylphosphine oxide, methylphosphonic acid dimethyl ester
12958
[methoxy(methyl)phosphoryl]oxymethane
COP(C)(=O)OC
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Normativo: Possono essere applicate restrizioni all'acquisto di questo prodotto e potremmo avere bisogno di richiedere ulteriore documentazione all'utente.
756-79-6
C3H9O3P
124.08
MFCD00008349
VONWDASPFIQPDY-UHFFFAOYSA-N
dimethyl methylphosphonate, dimethylmethylphosphonate, pyrol dmmp, dmmp, dimethyl methanephosphonate, fyrol dmmp, phosphonic acid, methyl-, dimethyl ester, methanephosphonic acid dimethyl ester, dimethoxymethylphosphine oxide, methylphosphonic acid dimethyl ester
12958
[methoxy(methyl)phosphoryl]oxymethane
COP(C)(=O)OC

Dimethyl methylphosphonate is used as a catalyst and a reagent in organic synthesis for the conversion of esters to ketophosphonates. It is an additive for the synthesis of unsaturated polyester resin which has flame retardant high phosphorous and UV-cured epoxy acrylate. It finds application as hydraulic fluids as well.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl methylphosphonate is used as a catalyst and a reagent in organic synthesis for the conversion of esters to ketophosphonates. It is an additive for the synthesis of unsaturated polyester resin which has flame retardant high phosphorous and UV-cured epoxy acrylate. It finds application as hydraulic fluids as well.

Solubility
Miscible with water, ethanol, ether, alcohol, benzene, acetone and carbon tetrachloride. Immiscible with heavy mineral oil.

Notes
Incompatible materials strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Dimethyl methylphosphonate
Densità 1.16
Punti di ebollizione 180°C to 181°C
Punto d'infiammabilità 68°C (154°F)
Indice di rifrazione 1.413
Quantità 10 g
Beilstein 878263
Indice di Merck 14,3391
Informazioni di solubilità Miscible with water,ethanol,ether,alcohol,benzene,acetone and carbon tetrachloride. Immiscible with heavy mineral oil.
Peso formulazione 124.08
Percent Purity 97%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'