Learn More
Lenz Laborglasintrumente Dimroth Condensers for Extractors with GL Threads
Codice prodotto. 10101602
Cambia vista
Click to view available options
Nessun giunto rastremato standard:
NS 29/32
NS 34/35
NS 60/46
NS 71/51
Dimensione della confezione:
Pezzo
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Nessun giunto rastremato standard Dimensione della confezione
10101602 NS 34/35 Pezzo
10530583 NS 29/32 Pezzo
10355042 NS 71/51 Pezzo
10782903 NS 60/46 Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 10101602 Fornitore Lenz Laborglasintrumente N. del fornitore 05246015
  • ONLINE EXCLUSIVE PRICE
    131.00€
    72.08€ / pezzi 1
    Risparmia 58.92€
    44% Sconto
Spedizione stimata: 28-12-2026
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Dimroth condensers for extractors
  • With GL threads Dimroth condensers for extractors
  • For condensing vapours
  • With NS cone
  • Water connection: GL 14 thread (with two threaded plastic olives)
  • Made of DURAN™ tubing
Specifica
Nessun giunto rastremato standard NS 34/35
Materiale Duran Glass