Learn More
Diphenylacetaldehyde, TRC
Specifica
Specifica
| Materiale o nome chimico | Diphenylacetaldehyde |
| CAS | 947-91-1 |
| Formula molecolare | C14H12O |
| Quantità | 2.5 g |
| Sinonimo | alpha-Phenylbenzeneacetaldehyde, Diphenyl-Acetaldehyde, 2,2-Diphenylacetaldehyde, 2,2-Diphenylethanal, NSC 21645, alpha,alpha-Diphenylacetaldehyde, alpha-Phenylbenzeneacetaldehyde |
| InChI Formula | InChI=1S/C14H12O/c15-11-14(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-11,14H |
| SMILES | O=CC(c1ccccc1)c2ccccc2 |
| IUPAC Name | 2,2-diphenylacetaldehyde |
| Molecular Weight (g/mol) | 196.24 |
| Peso formulazione | 196.0888 |