Learn More
Thermo Scientific™ Replacement Tips for UNIFILTER and UNIGUARD Direct-Connection Holders
Product Code. 10468444 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
UNIFILTER HPLC Filter Systems and UNIGUARD Guard Cartridge Holders
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10468444 UNIFILTER HPLC Filter Systems and UNIGUARD Guard Cartridge Holders Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10468444 Supplier Thermo Scientific™ Supplier No. 850RT
Spedizione stimata: 20-11-2026
Standard replacement tips for Thermo Scientific™ UNIGUARD™ and UNIFILTER™ Direct-Connection Holders.
Use the Thermo Scientific™ Replacement Tip as a standard component for UNIFILTER™ Direct-Connection HPLC Filter Systems or for UNIGUARD™ Direct-Connection Guard Cartridge Holders. The UNIGUARD Cartridge Holder connectors have an inner diameter that matches the analytical column inset and eliminates the need for extra fittings. The UNIFILTER Filter Systems are replaceable 0.5μm drop-in filters that enhance column lifetime and improve performance. A Filter System comprises the holder, filter and replacement tip.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) UNIFILTER HPLC Filter Systems and UNIGUARD Guard Cartridge Holders
Product Type Standard Replacement Tip, CPI