Learn More
Lenz Laborglasintrumente Distilling Receivers Acc. to Anschütz Thiele, Straight, with Cooling Jacket
Codice prodotto. 10000491
Cambia vista
Click to view available options
Capacità (sistema metrico):
50 mL
100 mL
Materiale:
Duran Glass
Duran Tubing, PTFE
Dimensione della confezione:
Pezzo
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema metrico) Materiale unitSize
10000491 50 mL Duran Glass Pezzo
10683622 100 mL Duran Glass Pezzo
10244531 50 mL Duran Tubing, PTFE Pezzo
10468212 100 mL Duran Tubing, PTFE Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 10000491 Fornitore Lenz Laborglasintrumente N. del fornitore 05305004
Aggiungi al carrello
Aggiungi al carrello
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Receivers with straight adapter
  • With cooling jacket, graduated, with socket and cone NS 29/32
  • Vacuum connection: thread GL18
  • NS stopcock with solid glass or PTFE plug
  • Made of DURAN™ tubing
Specifica
Materiale Duran Glass
Capacità (sistema metrico) 50 mL
Giunto rastremato standard Socket and Cone NS 29/32