Learn More
Lenz Laborglasintrumente Distilling Receivers Acc.to Bernauer with Flasks
Product Code. 10306361
Change view
Click to view available options
Standard Taper Joint:
Upper Socket 12/23 NS
Upper Socket 19/26 NS
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Unit Size
10306361 Upper Socket 12/23 NS Each
10326361 Upper Socket 19/26 NS Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10306361 Supplier Lenz Laborglasintrumente Supplier No. 05288001
  • ONLINE EXCLUSIVE PRICE
    157.00€
    102.99€ / pezzi 1
    Risparmia 54.01€
    34% Sconto
Spedizione stimata: 11-12-2026
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Distilling receivers acc. to Bernauer (modified)
  • With three cones NS 14/23 and three flasks
  • Vacuum connection with glass olive
  • Made of DURAN™ tubing
Specifications
Material Duran Glass
Capacity (Metric) 50 mL
Quantity 1 Ea.
Standard Taper Joint Upper Socket 12/23 NS