Descrizione
- The DK 18/26 Series digestion unit with aluminum heating block ensures high thermal homogeneity heating up to 450°C. The temperature of the block is constantly controlled by a microprocessor and is extremely stable, homogeneous and precise: ±0.5°C at all selectable temperatures.
- The DK 18/26 Series guarantees accuracy and repeatability. The DK 18/26 performs batch digestion of up to 18 samples with 100 mL test tubes - 26 mm dia., reducing the energy consumption (-13% compared to the previous series). Low energy consumption provides lower costs of analysis and limits CO2 release.
- The design of the DK series has been carefully engineered to allow laboratories to save valuable space under the hood.
- DK Digesters have very compact dimensions: the new aluminum heating block has been lowered and the vertical proportions allow to free precious space in the laboratory.
- The structure is made of stainless steel that has been specifically designed and tested to ensure strong resistance to chemicals and mechanical stress.
- The aluminum block protection grid provides enhanced heating performance and protects the heating block from splashes and chemicals.
- The bright digital display provides maximum visibility and easy-to-read information.
- The instrument is operated with 4 keys to navigate the menu options and program the digestions in a simple and fast way.
- Through the intuitive graphic display it is possible to set up to 20 methods with 4 temperature ramps.
- Acoustic signals inform the operator of the completion of the analysis facilitating the daily use of the digester.
- The inclination of the front panel protects it from chemical spillage at high temperatures.
- For application where the digestion process produces fumes, acid gases or corrosive substances, the below specific accessories are strongly recommended:
- JP Pump aspirates noxious fumes
- SMS Scrubber neutralizes all the fumes
Specifica
Specifica
| Display | Digital |
| Tensione | 230 V |
| Capacità (sistema imperiale) | 18 x 3.4 oz. |
| Capacità (sistema metrico) | 18 x 100 mL |
| Dimensioni (L x P x A) | 130 x 420 x 230 mm |
| Include | Instrument Only |
| Materiale | Stainless Steel, Epoxy Painted with Chemical Resistant Coating |
| No. of Places | 18 |
| Temperatura massima (sistema imperiale) | 842°F |
| Temperatura massima (sistema metrico) | 450°C |
| Vedi altri risultati |