Learn More
DL-Cysteine Hydrochloride Hydrate, TRC
Specifica
Specifica
| Materiale o nome chimico | DL-Cysteine Hydrochloride Hydrate |
| CAS | 96998-61-7 |
| Formula molecolare | C3H7NO2S .HCl .xH2O |
| Quantità | 10 g |
| Sinonimo | Cysteine Hydrochloride Hydrate |
| InChI Formula | InChI=1S/C3H7NO2S.ClH.H2O/c4-2(1-7)3(5)6;;/h2,7H,1,4H2,(H,5,6);1H;1H2 |
| SMILES | SCC(N)C(O)=O.Cl.[H2O] |
| IUPAC Name | cysteine hydrochloride hydrate |
| Molecular Weight (g/mol) | 175.64 |