Learn More
DL-o-Tyrosine, 98%, Thermo Scientific™
Product Code. 11495290
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495290 0.25 g 0.25g
11405300 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11495290 Supplier Thermo Scientific Alfa Aesar Supplier No. 044541.01
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2370-61-8
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00021725
InChI Key WRFPVMFCRNYQNR-UHFFFAOYNA-N
Synonym dl-o-tyrosine, 2-amino-3-2-hydroxyphenyl propanoic acid, o-tyrosine, 2-hydroxyphenylalanine, 2-hydroxy-dl-phenylalanine, ortho-tyrosine, 2-tyrosine, o-tyrosine, dl, dl-3-o-hydroxyphenyl alanine, 2-amino-3-2-hydroxyphenyl propionic acid
PubChem CID 91482
IUPAC Name 2-amino-3-(2-hydroxyphenyl)propanoic acid
SMILES NC(CC1=CC=CC=C1O)C(O)=O
Specifications
Melting Point ∼256°C (decomposition)
Beilstein 2805789
Solubility Information Soluble in water,and DMSO (Heated).
Quantity 0.25 g
Formula Weight 181.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material DL-o-Tyrosine