Learn More
Abcam DL-Serine Assay Kit
Codice prodotto. 32120221 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
32120221 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 32120221 Fornitore Abcam N. del fornitore ab241027100Test
 Articolo soggetto a restrizioni
Spedizione stimata: 25-11-2026
Measure D Serine in 1h 15m in Biofluids, Cell/Tissue Extracts with DL-Serine Assay Kit AB241027. Sensitivity <0.1 nmol/well. Cited in 8 publications. Use with Microplate reader.
Measure D Serine in 1h 15m in Biofluids, Cell/Tissue Extracts with DL-Serine Assay Kit AB241027. Sensitivity <0.1 nmol/well. Cited in 8 publications. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 100 Tests
Status giuridico RUO