Learn More
Thermo Scientific Chemicals DL-Valine, +99%
Catalog Number p-211489
Click to view available options
Quantità:
50 g
250 g
1 kg
Imballaggio:
Plastic Bottle
516-06-3
C5H11NO2
117.15
MFCD00004267
KZSNJWFQEVHDMF-UHFFFAOYSA-N
dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid
1182
CHEBI:27266
2-amino-3-methylbutanoic acid
CC(C)C(N)C(O)=O
516-06-3
C5H11NO2
117.15
MFCD00004267
KZSNJWFQEVHDMF-UHFFFAOYSA-N
dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid
1182
CHEBI:27266
2-amino-3-methylbutanoic acid
CC(C)C(N)C(O)=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
TRUSTED_SUSTAINABILITY
Materiale o nome chimico DL-Valine
Punto di fusione 283.5°C to 285.0°C
Nota nome 99+%
Colore White
Cloro (Cl) 400ppm max.
Metalli pesanti (come Pb) 10ppm max.
Intervallo percentuale saggio 99+%
Spettro a infrarossi Authentic
Formula lineare (CH3)2CHCH(NH2)CO2H
Beilstein 04, 430
Perdita all'essiccatura 0.2% max. (105°C, 3 hrs)
Indice di Merck 14, 9909
Imballaggio Plastic Bottle
Informazioni di solubilità Solubility in water: 68g/L. Other solubilities: practically insoluble in cold alcohol and ether, soluble in mineral acids, insoluble in organic solvents, slightly soluble in benzene, ethanol
Peso formulazione 117.15
Trasmittanza 95% min. (c=10, 2N HCl, 430nm) 1cm cell
Percent Purity ≥99%
Cenere solfata 0.2% max.
Grado Biochemical
Densità 1.3100g/mL
Quantità 50 g
Gravità specifica 1.31
Forma fisica Crystalline Powder