missing translation for 'onlineSavingsMsg'
Learn More

DNAJB2 Antibody (2H8) [mFluor Violet 610 SE], Novus Biologicals Biologicals™

Codice prodotto. 30519241 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
30519241 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30519241 Fornitore Novus Biologicals N. del fornitore NBP342958MFV610

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DNAJB2 Monoclonal antibody specifically detects DNAJB2 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigene DNAJB2
Applicazioni ELISA, Western Blot
Classificazione Monoclonal
Clone 2H8
Coniugato mFluor Violet 610 SE
Formulazione 50mM Sodium Borate
Alias gene DnaJ (Hsp40) homolog, subfamily B, member 2, HSJ-1, HSPF3neuronal DNAJ-like 1
Specie ospite Mouse
Immunogeno DnaJ(Hsp40) homolog, subfamily B, member 2 - (Uniprot# P25686)
Metodo di purificazione Protein A or G purified
Quantità 0.1 mL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 3300
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Forma Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.