Learn More
Thermo Scientific™ Domed PCR Caps, strips of 12
Product Code. 11594774
Change view
Click to view available options
Quantity:
10 packs of 8
25 packs of 8
Color:
Clear
Unit Size:
Pack of 200
Pack of 80
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Color Unit Size
11594774 25 packs of 8 Clear Pack of 200
11584774 10 packs of 8 Clear Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11594774 Supplier Thermo Scientific™ Supplier No. AB0853
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In stock
Add to basket
Add to basket
Bulk Quote Request
Domed cap strips for PCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights:
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
Certifications:
Certified free from human DNA, RNases, and DNases.
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Polypropylene
No. of Wells 12
Quantity 25 packs of 8
Format Strips of 12 Domed Caps
Product Type PCR Plate Cap Strip
Certified RNase/DNase-free Yes
Plate Compatibility 96-well Plate