Learn More
Emma Safety Footwear Donovan (Ruffneck) Safety Shoes
Looks and feels like a classic sneaker. Highly supportive reinforced ruff neck heel together with the shock absorbing sole keeps ankles, shins and joints free from stress and injury.
Marca:  Emma Safety Footwear 974546-48
Dettagli aggiuntivi : Peso : 1.98800kg
 vedi altre versioni di questo prodotto
Codice prodotto. 15939131
Quantità Prezzo
1 101.59€ / 1 paio
10 95.64€ / 1 paio
Spedizione stimata
20-01-2021
Aggiungi al carrello
Descrizione
Descrizione
  • S3-Safety shoe with sole plate for wet work
  • Nubuck / Oil Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • ESD certified
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifica
Specifica
EN ISO 20345:2011, EN 12568:2010
Brown
Hydro-Tec™ Sanitized Silver
48
Laces
Donovan (Ruffneck)
Nubuck, Oiled Nubuck
Steel