Learn More
Metro™ Double-Deep qwikTRAK™ Mobile Unit Kits
Kit provides components for shelving units that fit on tracks for added mobility
Marca:  Metro™ LBTM24X3
Dettagli aggiuntivi : Peso : 13.06400kg
Codice prodotto. 15946991
  • 2230.00€ / Pezzo
Spedizione stimata: 18-09-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • For use with Metro Super Erecta™, Super Erecta Pro™ Super Adjustable™ and MetroMax i™ shelving systems
  • Kit includes eight 74 in. (188cm) high -UP posts, four V-groove casters; one stainless steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Order Info
Order one kit per double-deep system. Shelves are sold separately.
Specifica
Specifica
Eight 74 in. high up posts, four V-groove casters, one Stainless Steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
24 in.
61 cm
Heavy Polymer
Polymer
24 in.
Mobile Unit Kit
Metro™ MetroMax i™ shelving system
61 cm