missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxylamine N-Oxide, TRC
Specifica
Specifica
| Materiale o nome chimico | Doxylamine N-Oxide |
| CAS | 97143-65-2 |
| Note sul grado di purezza | HPLC |
| Formula molecolare | C17 H22 N2 O2 |
| Quantità | 25 mg |
| Sinonimo | Ethanamine, N,N-dimethyl-2-[1-phenyl-1-(2-pyridinyl)ethoxy]-, N-oxide (9CI, ACI), Doxylamine N-oxide, N,N-Dimethyl-2-[1-phenyl-1-(2-pyridinyl)ethoxy]ethanamine N-oxide, Doxylamine N-oxide (aliphatic N-oxide) |
| InChI Formula | InChI=1S/C17H22N2O2/c1-17(15-9-5-4-6-10-15,16-11-7-8-12-18-16)21-14-13-19(2,3)20/h4-12H,13-14H2,1-3H3 |
| SMILES | CC(OCC[N+](C)(C)[O-])(c1ccccc1)c2ccccn2 |
| IUPAC Name | N,N-dimethyl-2-(1-phenyl-1-pyridin-2-ylethoxy)ethanamine oxide |
| Molecular Weight (g/mol) | 286.369 |
| Vedi altri risultati |
Titolo del prodotto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?