Learn More
Abnova™ DPYD/CEN1p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Marca: Abnova™ FG0219.200uL
Dettagli aggiuntivi : Peso : 0.02000kg
Specifica
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
DHP,DHPDHASE,DPD,MGC132008,MGC70799 | |
1806 | |
200 μL |
Tube | |
dihydropyrimidine dehydrogenase | |
DPYD | |
DNA/RNA | |
Liquid |