Learn More
Dräger™ CMS Chips: Nitrogen Dioxide
Product Code. 10641733
Change view
Click to view available options
Measurement Range:
0.5 to 25
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
10641733 0.5 to 25 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10641733 Supplier Dräger™ Supplier No. 6406120
  • ONLINE EXCLUSIVE PRICE
    250.28€
    202.90€ / pezzi 1
    Risparmia 47.38€
    18% Sconto
Spedizione stimata: 04-12-2026
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Nitrogen Dioxide CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 0.5 to 25
Response Time 20 to 330 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C