Learn More
Dräger™ Short-Term Detector Tubes - Hydrogen Sulfide
Product Code. 10276821
Change view
Click to view available options
Components Measured:
Hydrogen Sulfide 0.5/a
Hydrogen Sulfide 1/d
Hydrogen Sulfide 100/a
Hydrogen Sulfide 5/b
Measurement Range:
0.5 to 15ppm / 5 to 150ppm
1 to 200ppm
100 to 2000ppm
5 to 60ppm
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured Measurement Range Unit Size
10276821 Hydrogen Sulfide 100/a 100 to 2000ppm Pack of 10
10558741 Hydrogen Sulfide 1/d 1 to 200ppm Pack of 10
10142032 Hydrogen Sulfide 0.5/a 0.5 to 15ppm / 5 to 150ppm Pack of 10
10378421 Hydrogen Sulfide 5/b 5 to 60ppm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10276821 Supplier Dräger™ Supplier No. CH29101
  • ONLINE EXCLUSIVE PRICE
    188.51€
    146.21€ / pezzi 10
    Risparmia 42.30€
    22% Sconto
 Restricted Item
Spedizione stimata: 03-12-2026
Cost-effective and reliable detector tube for the measurement of hydrogen sulfide gas.
Dräger short-term detector tubes for spot gas measurements.
  • Target Gas: Hydrogen Sulfide (H2S)
  • Measurement Range: 100 to 2000 ppm
  • Number of Strokes (n): 1
  • Measurement Duration: ca. 30 sec.
  • Color Change: White to brown
  • Standard Deviation: 5% to 10%
  • Operating Temperature: 0°C to 40°C
  • Operating Humidity: 3 to 40 mg H2O/L
  • Reaction Principle: H2S + Pb2+ → PbS + 2H+
  • Cross Sensitivity: 2000 ppm Sulfur dioxide and 100 ppm Nitrogen dioxide do not interfere.
  • Compatibility: For use with Dräger Accuro™ bellows pump, Accuro 2000, or Quantimeter 1000.
Specifications
Components Measured Hydrogen Sulfide 100/a
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Humidity Range 3 to 40mg/L
Measurement Range 100 to 2000ppm
No. of Strokes 1
Quantity 10
Response Time 30 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C