Learn More
BRAND™ SILBERBRAND™ AR-GLAS™ Spare Burette
Product Code. 10647671 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
10 mL
15 mL
50 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10647671 50 mL Each
10238460 10 mL Each
10064310 15 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10647671 Supplier BRAND™ Supplier No. 23718
 Restricted Item
Spedizione stimata: 16-12-2026
Manufactured with Dr. Schilling pattern from AR-GLAS™. For use with Automatic Burettes.
  • AR-GLAS™ construction
  • For use with automatic burette
  • Dr. Schilling pattern
  • Schellbach style back
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50 mL
Length (Metric) 580 mm
For Use With (Equipment) Automatic burettes
Material Borosilicate Glass
Type Automatic Buret