Learn More
Thermo Scientific™ DRI™ Creatinine-Detect™ Specimen Validity Test
Codice prodotto. 17516015 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Quantità:
500 mL
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
17516015 500 mL Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 17516015 Fornitore Thermo Scientific™ N. del fornitore 1797
 Articolo soggetto a restrizioni
Spedizione stimata: 10-12-2026
Rise to the challenge of combating the use of additives and adulterants to mask controlled substances with the Thermo Scientific™ DRI™ Creatinine-Detect™ Specimen Validity Test.
Combat the use of additives and adulterants to mask controlled substances in urine samples with Thermo Scientific™ DRI™ Creatinine-Detect™ Specimen Validity Tests. Liquid and ready-to-use, these tests can be performed on a variety of general chemistry analyzers.
  • Associated calibrators and controls offered in convenient sizes
  • Complete and automated way to test for sample integrity and adulteration
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage Room Temperature
Descrizione DRI Creatinine-Detect Assay Specimen Validity Test
Analiti rilevabili Creatinine
Tipo di campione Urine
Tipo Reagent
Quantità 500 mL