Learn More
Fisherbrand™ Drosophila Products, BuzzPlugs™ Vial and Bottle Closures
Sfoglia Tutto Fisherbrand Traceable ProdottiDescrizione
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
- Superior barrier to mite infestation
- Exact fit with narrow 0.9 in. (25mm) or wide 1.1 in. (28.5mm) dia. plastic shell vials or standard (≈24mm dia.) glass vials or plastic stock bottles
- Stretches yet maintains tight fit after repeated removal and closing
- Flat surface accepts marking pen
Specifica
Specifica
| Da utilizzare con (applicazione) | Minimizes evaporation, keeps food fresh longer |
| Da utilizzare con (apparecchiatura) | 2.5cm narrow plastic shell vials |
| Include | Superior barrier |
| Quantità | Scatola da 1000 |
| Materiale | Cellulose Acetate |
| Tipo | BuzzPlugs Bottle Closure |