Learn More
DWK Life Sciences DURAN™ Acid Storage Bottle, Amber, Includes Standard Taper Pennyhead stopper, Complete with ground joint glass cap
Catalog Number p-7189298
Click to view available options
Capacità (sistema metrico):
100 mL
250 mL
500 mL
1000 mL
DURAN™ amber glass acid storage bottle.
Made of chemically resistant amber DURAN™ glass. With standard taper pennyhead stopper and additional ground glass cap. This provides an improved seal and protection against acid vapours. Ambered glass provide protection from Ultra-violet light.
  • Protection from UV light due to amber colouration
  • Ground DURAN™ glass stopper and additional DURAN™ glass cap provides protection against acid vapours
  • Pennyhead stopper with standard taper joint
  • Conical shoulders
  • Replacable glass cap
Diametro (sistema metrico) 75 mm
Capacità (sistema metrico) 250 mL
Da utilizzare con (applicazione) Storage and dispensing of acids; Storage of photosensitive reagents
Materiale Borosilicate Glass (3.3)
Quantità 10/Pk.