missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Acid Storage Bottle, Amber, Includes Standard Taper Pennyhead stopper, Complete with ground joint glass cap
Click to view available options
Capacità (sistema metrico):
100 mL
250 mL
500 mL
1000 mL
Descrizione
Made of chemically resistant amber DURAN™ glass. With standard taper pennyhead stopper and additional ground glass cap. This provides an improved seal and protection against acid vapours. Ambered glass provide protection from Ultra-violet light.
- Protection from UV light due to amber colouration
- Ground DURAN™ glass stopper and additional DURAN™ glass cap provides protection against acid vapours
- Pennyhead stopper with standard taper joint
- Conical shoulders
- Replacable glass cap
Specifica
Diametro (sistema metrico) | 109 mm |
Capacità (sistema metrico) | 1000 mL |
Da utilizzare con (applicazione) | Storage and dispensing of acids; Storage of photosensitive reagents |
Materiale | Borosilicate Glass (3.3) |
Quantità | 10/Pk. |
Autoclavabile | Autoclavable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'